Learn More
Glutaraldehyde, 25% aq. soln.
Description
Glutaraldehyde is used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues and as an amine-reactive homobifunctional crosslinker. It is also used in coupling reaction of biotin hydrazides to nucleic acids which is useful for conjugating fluorescent hydrazides and hydroxylamines to DNA and for stabilization ofproteins on agarose beads,activation of polystyrene and glass, for immobilization of antibodies and antigens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Glutaraldehyde is used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues and as an amine-reactive homobifunctional crosslinker and as a disinfectant for medical equipment. It is also used in coupling reaction of biotin hydrazides to nucleic acids which is useful for conjugating fluorescent hydrazides and hydroxylamines to DNA and for stabilization ofproteins on agarose beads,activation of polystyrene and glass, for immobilization of antibodies and antigens. It is a disinfectant, which is rapidly effective against vegetative forms of Gram-positiveand Gram-negative bacteria.
Solubility
Miscible with water, ethanol, benzene, ether, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.
Notes
Heat and air sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents.
Specifications
Specifications
| Density | 1.062 |
| Assay Percent Range | 0.25 |
| Refractive Index | 1.373 |
| Linear Formula | OHC(CH2)3CHO |
| Quantity | 2500 mL |
| UN Number | UN2922 |
| Beilstein | 605390 |
| Merck Index | 14,4472 |
| Solubility Information | Miscible with water,ethanol,benzene,ether,acetone,dichloromethane,ethylacetate,isopropanol,n-hexane and toluene. |
| Formula Weight | 100.12 |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H314-H317-H334 |
| Hazard Statement | GHS H Statement H331-H334-H314-H302-H317 Toxic if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 203-856-5 |
| RTECSNumber | MA2450000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.