Learn More
Ted Pella Inc Glutaraldehyde, 111-30-8, MFCD00007025, 70% EM grade, 10 x 2ml
SDP
Supplier:  Ted Pella Inc 18436
Encompass
Linear Formula: OHC(CH2)3CHO, Molecular Weight: 100.12, Synonym: Glutaric dialdehyde solution, Pentane-1,5-dial. Glutaraldehyde solution has been used as a fixative for transfected adherent monkey kidney fibroblasts (COS-7) cells for electron microscopy studies, human osteosarcoma (U2OS) cells for super-resolution (SR) experiments and for hydrogels for scanning electron microscopy (SEM) studies.
Catalog No. NC9960165
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.