Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaronitrile 96.0+%, TCI America™
SDP

Catalog No. G0072250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0072250G 250 g
G007225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0072250G Supplier TCI America Supplier No. G0072250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 544-13-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00001970
InChI Key ZTOMUSMDRMJOTH-UHFFFAOYSA-N
Synonym glutaronitrile, 1,3-dicyanopropane, glutarodinitrile, glutaric acid dinitrile, 1,3-trimethylenedinitrile, pyrotartaric acid nitrile, glutaronitrle, unii-01zi68f3cq, trimethylene dicyanide, glutaric dinitrile
PubChem CID 10994
IUPAC Name pentanedinitrile
SMILES C(CC#N)CC#N

Specifications

Color Yellow
Boiling Point 286°C
Quantity 250 g
UN Number 3276
Formula Weight 94.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Glutaronitrile
RTECSNumber YI3500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.