Learn More
Cayman Chemical Glutathione Assay Kit, 480 wells
SDP
Supplier:  Cayman Chemical 703002480WELLS
Encompass_Preferred
Cayman’s GSH Assay utilizes a carefully optimized enzymatic recycling method for the quantification of GSH in a 96-well microplate format. It measures both GSH and GSSG to reflect total glutathione in a sample. The kit can also be used to measure only GSSG by following an alternative protocol. The GSH Assay can be used for plasma, tissue samples, and cultured cells with minimal sample processing.
Catalog No. NC0039674