Learn More
XpressBio Xpress Capture Glutathione ELISA Plates, Black 384 wells (5-pack)
SDP
Supplier:  XpressBio EX7000B384
Encompass_Preferred
Capture coated Glutathione ELISA plates may be used with GST-tagged proteins for several uses including recombinant protein expression screening, immunoabsorbtion assays, biochemical assays, competition assays and protein purification.
Catalog No. 50-171-7645
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.