Learn More
Arbor Assays Glutathione Fluorescent 384-Well Detection Kit (2 Plate)
SDP
Supplier:  Arbor Assays K006F1D
Encompass_Preferred
The DetectX Glutathione (GSH) Fluorescent Detection Kit utilizes a proprietary non-fluorescent molecule, ThioStar, that covalently binds to the free thiol group on GSH to yield a highly fluorescent product.
Catalog No. 50-227-8384
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.