Learn More
                                    Bachem H-Glu(Cys-Gly-OEt)-OEt
Supplier: Bachem 4025802.0250
            Glutathione-diethyl ester (reduced), H-Glu(Cys-Gly-OEt)-OEt, 97451-40-6, C14H25N3O6S, Mr 363.44, 250mg. Human cells transport glutathione diethyl ester much more effectively than the corresponding monoethyl ester. Hence, the diethyl ester of glutathione could be used for decreasing oxidative stress and toxicity.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.