Learn More
Cayman Chemical Glutathione ethyl ester, 92614-59-0, 50 mg
SDP
Supplier:  Cayman Chemical 1495350
Encompass_Preferred
Molecular Formula C12H21N3O6S, Purity ≥95%, Formula Weight 335.4, Synonyms: GSH ethyl ester; A cell permeable derivative of GSH that undergoes hydrolysis by intracellular esterases to release GSH; used to protect cells against damage from radiation, oxidants, and various toxic compounds including heavy metals.
Catalog No. NC0861733
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.