Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glutathione Monoclonal Antibody (D8)

Catalog No. 01670354
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-354 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-354 Supplier Invitrogen™ Supplier No. MA17620
  • $660.50
    $498.40 / Each of 1

    Save $162.10

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-7620 detects Glutathione from all organism samples. MA1-7620 has been successfully used in ELISA, WB and IP procedures. The MA1-7620 immunogen is a Glutathione protein. Store at 4°C short term. For long term storage, store at -80°C.

Glutathione is a tripeptide comprised of three amino acids (cysteine, glutamic acid, and glycine) present in most mammalian tissue. Glutathione acts as an antioxidant, a free radical scavenger and a detoxifying agent. Glutathione is also important as a cofactor for the enzyme glutathione peroxidase, in the uptake of amino acids, and in the synthesis of leukotrienes. As a substrate for glutathione S-transferase, this agent reacts with a number of harmful chemical species, such as halides, epoxides and free radicals, to form harmless inactive products. In erythrocytes, these reactions prevent oxidative damage through the reduction of methemoglobin and peroxides. Glutathione is also involved in the formation and maintenance of disulfide bonds in proteins and in the transport of amino acids across cell membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutathione
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone D8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias C10H17N3O6S; Glutathion; Glutathione-SH; GPX4; L-Glutathione; Reduced glutathione
Host Species Mouse
Immunogen Glutathione.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.