Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Glutathione Sepharose™ High Performance Media

Catalog No. 45000331 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-331 100 mL
45-000-330 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-331 Supplier Cytiva Supplier No. 17527902
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For high resolution purification of Glutathione S transferase (GST) tagged proteins

  • One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
  • GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione
  • Mild purification process preserves protein antigenicity and function
  • Glutathione ligand is coupled to 6% agarose matrix

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity >Approx. 10 mg GST-tagged protein/mL resin
Description Glutathione Sepharose High Performance is an affinity resin recommended for purifying glutathione S-transferase (GST)-tagged proteins when high resolution is the priority.
Flow Rate <600 cm/hr.
Kit Contents 6% Agarose, Binding capacity ∼10mg recombinant GST/mL media
Matrix Highly crossed-linked agarose, 6%
pH Range 3 to 12
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4°C to 30°C, 20% Ethanol
Particle Size 34 μm
Quantity 100 mL
Type Chromatography Media
Ligand Type Glutathione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.