Learn More
Penn Veterinary Supply Inc GLUTURE 1.5ML
SDP
Supplier:  Penn Veterinary Supply Inc ABT3204604
Encompass
GLUTURE Topical Tissue Adhesive - ABBOTT LABS. 1.5ml tube. -Sets up quickly and creates a strong bond -Flexible - contours to wound site -Applies purple for excellent visibility and dries clear for a neat appearance -Precise delivery - pipette applicator aids placement control -Viscosity and pipette design enable flow controlled application -10 disposable pipette applicators in an economical 1.5 ml multi-use package, no refrigeration necessary
Catalog No. NC9855218
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.