Learn More
MWI VETERINARY GLUTURE TOPICAL ADHESIVE 1.5ML
Supplier: MWI VETERINARY 034207
Create a strong and flexible bond over wounds to assist healing with this topical tissue glue * Flexible, medical-grade tissue glue sets up quickly and contours to pet wound site * Pipette applicator for precision closure of wounds Aid natural healing of pet wounds with Gluture professional tissue glue. Gluture Topical Tissue Adhesive is a medical-grade, cyanoacrylate formulation that creates a strong AND flexible bond between wound sites. Gluture forms a thin, waterproof bridge/bandage that sets up quickly and contours to wound site. Applies purple for excellent visibility and then dries clear. Topical adhesive naturally sloughs off as the incision heals. Includes 10 disposable, precision-tip pipette applicators for convenient multiple use. 1.5 ml. Please Note: this product is not intended for infected or deep puncture wounds or as a replacement for suture material that would normally be used subcutaneously.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.