Learn More
ORS Nasco GLV 11INMD WT BUTYL SZ10 1PR
Supplier:  ORS Nasco 068B161/10
Encompass_Preferred
Honeywell part# B161/10. Honeywell North 16 mil Butyl Chemical Resistant Gloves Size: 10 Grip Design: Smooth Black Glove Cuff Style: Beaded/Rolled Latex Free: Yes Style: North Butyl B161 Thickness: 16 mil 0.40 mm Certifications/Compliance: ASTM D3389-05 ASTM F790-97 OSHA 29 CFR EN 374 EN 420 Hand: Specific
Catalog No. 50-255-1639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.