Learn More
ORS Nasco GLV BUTYL 13ML SZ 9 1PR
Supplier:  ORS Nasco 068B131/9
Encompass_Preferred
Honeywell part# B131R/10. Honeywell North 13 mil Butyl Chemical Resistant Gloves Black Glove Cuff Style: Beaded/Rolled Latex Free: Yes Size: 10 Style: North Butyl B131 Thickness: 13 mil 0.33 mm Certifications/Compliance: ASTM D3389-05 ASTM F790-97 OSHA 29 CFR EN 374 EN 420 EN388 Standard: EN 388 Hand: Specific
Catalog No. 50-255-1646
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.