Learn More
ORS Nasco GLV DRY BOX NEOP 15 MIL 1PR
Supplier:  ORS Nasco 0688N1532/9Q
Encompass_Preferred
Honeywell part# 8N1532/9Q. Honeywell Neoprene Glove Box Gloves 8 in. Port Black Hand: Hand specific Latex Free: Yes Port Size: 20.3 cm 8 in. Size: 9.75 Thickness: 15 mil 0.38 mm Certifications/Compliance: EN 388:2016 EN 374-3 Circumference Hand: 9.75 in. 25 cm Diameter Cuff: 8 in. 20.3 cm Grip Design: Smooth
Catalog No. 50-255-1655
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.