Learn More
Abcam Gly-Phe beta-naphthylamide, Cathepsin C substrate, 1G
Supplier:  Abcam AB1459141G
Encompass_Preferred
MW 347.41 g/mol, Purity >95%. Substrate for the lysosomal cysteine protease Cathepsin C. Accumulates within the lysosome. Hydrolysis by cathepsin C degrades Gly-Phe-β-naphthylamide into fragments that do not easily diffuse through the lysosomal membrane, leading to a loss of lysosome membrane integrity. Used to study intralysosomal hydrolysis, lysosomal membrane permeability, and the function of cathepsin C. Inhibits the cathepsin-dependent activation of caspase-8.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5016
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.