Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories Glycated Serum Protein Control

Catalog No. 23700558
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-700-558 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-700-558 Supplier Diazyme Laboratories Supplier No. DZ112BCON
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the testing of Glycated Serum Protein (Fructosamine) in human serum.

  • For glycated serum protein provides improved specificity and reliability compared to conventional NBT-based methods
  • The assay is highly precise with a total CV ≤ 1.3%
  • The assay offers extensive linearity up to 1354 μmol/L
  • The assay has no significant interference from: ascorbic acid (5 mg /dL); bilirubin (7.5 mg/dL); bilirubin conjugated (5 mg/dL); glucose (2400 mg/dL; hemoglobin (200 mg/dL); triglycerides (2000 mg/dL); and uric acid (35 mg/dL)
  • The measurement of glycated serum proteins is useful for monitoring diabetic patients
  • For in vitro diagnostic use only

Specifications

Certifications/Compliance CE, 510(k) Cleared, Health Canada Registered
Content And Storage Store at 2°C to 8°C. Do Not Freeze. Store CONTROL tightly capped when not in use.
Description Glkycated Serum Control Kit
Format 2 x 1 mL Vial
Type Diagnostic
Includes The Diazyme GSP Control Kit consists of two vials.
Quantity 2 Vials
Stability Unconstituted material: stability is up to the expiration date on label when stored at 2 to 8°C. Constituted material: stability is up to 14 days when stored at 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.