Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Glycerin
SCI_ED

Catalog No. S25342B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
1 L
4 L
Packaging:
ACS Grade - Liquid
Lab Grade - Liquid
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25342B 1 L Lab Grade - Liquid
S25342A 500 mL ACS Grade - Liquid
S25342D 4 L ACS Grade - Liquid
S25342C 4 L Lab Grade - Liquid
S25342 50 mL Lab Grade - Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S25342B
Add to Cart
Add to Cart

High-purity glycerin for classroom and lab use. Ideal for chemistry, biology, and physical science experiments.

  • Product Type: Glycerin (Glycerol)
  • Chemical Name or Material: Glycerin
  • Grade: ACS
  • Assay Percent Range: 99–100% w/v
  • Density: 1.2610 g/cm³ at 20°C
  • Physical Form: Liquid
  • Quantity: 1 L
  • Packaging: Lab Grade
  • Formula: C3H5(OH)3
  • Formula Weight: 92.1
  • Intended Use: For educational laboratory applications only (no Certificate of Analysis available)

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.2610g/°Cm3 at 20°C
Assay Percent Range 99 to 100% w/v
Packaging Lab Grade - Liquid
Linear Formula C3H5(OH)3
Quantity 1 L
Formula Weight 92.1
Grade ACS
Physical Form Liquid
Chemical Name or Material Glycerin
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.