Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, BiotechGrade, Approx. 99%, Spectrum™ Chemical
SDP

Catalog No. 18602950 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-950 500 mL Poly Bottle
18-602-951 4 L Poly Bottle
18-602-952 20 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-950 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G1433500ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

G1433, 56-81-5, C3H8O3

Spectrum™ Chemical Glycerin, BiotechGrade , is a sugar alcohol (polyol) compound. An odorless, viscous and colorless liquid that has three hydroxyl groups that allow its solubility in water and give it it's hygroscopic nature. Used in the personal care and pharmaceutical industry for improving smoothness and lubrication it is also used as a solvent and preservative.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Max % 100%
Boiling Point 290°C (554°F)
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 500 mL
Percent Purity ∼99%
Grade Biotech
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Glycerin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.