Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, Natural, EP, BP, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18602959 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Glass Bottle
Poly Bottle
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-959 500 mL Glass Bottle
18-602-958 500 mL Poly Bottle
18-602-968 1 L Poly Bottle
18-602-960 6 x 500 mL Glass Bottle
18-602-961 6 x 500 mL Poly Bottle
18-602-963 4 L Poly Bottle
18-602-964 4 x 4 L Glass Bottle
18-602-965 4 x 4 L Poly Bottle
18-602-966 20 L Poly Bottle
18-602-967 200 L Poly Bottle
18-602-962 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-602-959 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G1016500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

G1016, 56-81-5, C3H8O3

Spectrum™ Chemical Glycerin, Natural, USP, EP, BP is one of two USP grade Glycerins available from Spectrum with the other being Glycerin, Synthetic, USP, EP, BP, JP. Natural glycerin is produced from the hydrolysis of animal or vegetable lipids during soap manufacture but the majority of natural glycerin now comes as a result of it being a side product during the production of biodiesel. All Spectrum Chemical USP, EP, BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Max % 100%
Boiling Point 290°C (554°F)
Ignition Residue 0.01%
Packaging Glass Bottle
Refractive Index 1.470-1.475
Quantity 500 mL
Percent Purity 99 to 101%
Grade Multi-Compendial
Physical Form Liquid
Chemical Name or Material Glycerin, Natural
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.