Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, Synthetic, EP, BP, JP, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18602984 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 x 4 L
Packaging:
Amber Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-984 4 x 4 L Amber Glass Bottle
18-602-980 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-984 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G10154LTGLCS4

missing translation for 'validMainframeMsg'

G1015, 56-81-5, C3H8O3

Spectrum™ Chemical Glycerin, Synthetic, USP, EP, BP, JP is one of two USP grade Glycerins available from Spectrum with the other being Glycerin, Natural, USP, EP, BP. For those products requiring FDA approvals, if the initial formulation was done with Glycerin, Synthetic, USP, EP, BP, JP, the use of Glycerin, Natural, USP, EP, BP, JP is not interchangeable but would require the new formulation to undergo a new FDA approval process.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Max % 100%
Ignition Residue 0.01%
Packaging Amber Glass Bottle
Refractive Index 1.470-1.475
Quantity 4 x 4 L
Specific Gravity 1.258
Percent Purity 99 to 101%
Grade Multi Compendia
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.