Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol, 85% (w/w), Ricca Chemical
SDP

Catalog No. 3291816 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32918-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32918-16 Supplier Ricca Chemical Company Supplier No. 3291.816
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56-81-5 , 7732-18-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) Mixture
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Min % 84.0, 14.0
CAS Max % 86.0, 16.0
Color Colorless
Packaging Natural Poly Bottle
Quantity 500 mL
Percent Purity 85% w/w
Grade Laboratory
Concentration or Composition (by Analyte or Components) 85% (w/w) Aqueous Solution
Physical Form Liquid
Chemical Name or Material Glycerol
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.