Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol, ACS Reagent, ≥99.5%, Solstice

Catalog No. 60048027 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2 L
4 L
4 x 4 L
Packaging:
Glass bottle
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-027 4 x 4 L Glass bottle
60-048-026 4 L Poly bottle
60-048-025 2 L Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-048-027 Supplier Solstice Supplier No. G78934X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.25g/mL
pH Neutral
Boiling Point 290°C (1013 hPa)
Flash Point 160°C (320°F)
Ignition Point 698°F
Odor Odorless
Packaging Glass bottle
Linear Formula HOCH2CH(OH)CH2OH
Quantity 4 x 4 L
UN Number NONH for all modes of transport
Beilstein 635685
Solubility Information Completely miscible in water
Vapor Density 3.1 (vs air)
Vapor Pressure 1mmHg (20°C)
Formula Weight 92.09g/mol
Percent Purity ≥99.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less
RTECSNumber 200-289-5
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.