Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol, 99.5%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. AC327255000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC327255000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC327255000 Supplier Thermo Scientific Chemicals Supplier No. 327255000
  • $133.00
    $105.65 / Each of 1

    Save $27.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Absorbance (0.5M in water), (0.5 M in water) at 0nm, 0.02 max. at 280nm, 0.06 max. at 260nm
Melting Point 18.0°C
Density 1.2610g/mL
pH 6 to 7 (10% soln. at 25°C)
Boiling Point 290.0°C
Flash Point 160°C
Assay Percent Range 99.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4716
Linear Formula HOCH2CH(OH)CH2OH
Quantity 500 mL
Beilstein 01, 502
Specific Gravity 1.261
Merck Index 15, 4520
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
Viscosity 1069 mPa.s (20°C)
Formula Weight 92.09
Percent Purity 99.5%
Grade Molecular Biology
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-289-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.