Learn More
Glycerol, ACS, 99.5+%
Description
Glycerol is widely used in cosmetics, especially for production of soaps and lotions, solvents and emulsifiers. It may also be found in inks, adhesives and pharmaceuticals. It plays an important role in the manufacture of plastics and resins. Glycerol bonded with silica gel plays an important role for effective removal of traces of Co(II) from aqueous solutions. Also useful for optical clearing of gastric tissue.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 18°C |
| Density | 1.26 g/cm3 |
| Boiling Point | 182°C (20 mm) |
| Flash Point | 160°C (320°F) |
| Ignition Point | 400°C (752°F) |
| Assay Percent Range | >99.5% |
| Refractive Index | 1.474 |
| Quantity | 500 mL |
| Beilstein | 635685 |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H319: Causes serious eye irritation. |
| EINECSNumber | 200-289-5 |
| RTECSNumber | MA8050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.