Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol ACS AR, Macron™

Catalog No. MK5092200 Shop All Avantor Products
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-5092-200 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-5092-200 Supplier Avantor Supplier No. 509226

Specifications

CAS 56-81-5
Packaging Poly Drum
Molecular Formula C3H8O3
Quantity 200 L
MDL Number MFCD00004722
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES OCC(O)CO
IUPAC Name propane-1,2,3-triol
Molecular Weight (g/mol) 92.09
PubChem CID 753
ChEBI CHEBI:17754
Formula Weight 92.1
Grade ACS
Chemical Name or Material Glycerol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.