Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycerol-d8 | 7325-17-9 | MFCD00084149 | 98.3% | 100.14 g/mol | C3D8O3 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW127572100MG

Encompass_Preferred

Glycerol-d8 is the deuterium-labeled form of glycerol (C3D8O3) intended for research use as an internal standard and tracer in analytical chemistry. It is supplied with high isotopic enrichment and a stated purity for quantitative analyses by spectrometric and chromatographic techniques.

  • Deuterium-labeled glycerol suitable as an internal standard.
  • High isotopic enrichment (d8) for clear mass and spectral separation.
  • Purity: 98.3%.
  • Molecular weight: 100.14 g/mol.
  • Common applications: quantitative analysis by NMR, GC-MS, and LC-MS.
  • For research use only; not for therapeutic or clinical use.

Catalog No. 50-002-83101


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.