Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol Dimethacrylate (mixture of 1,2- and 1,3-form) (stabilized with MEHQ) 90.0+%, TCI America™
SDP

Catalog No. B293825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B293825G 25 g
B2938500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B293825G Supplier TCI America Supplier No. B293825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1830-78-0
Molecular Formula C11H16O5
Molecular Weight (g/mol) 228.24
MDL Number MFCD00066299
InChI Key OQHMGFSAURFQAF-UHFFFAOYSA-N
Synonym Bis(methacryloyloxy)propanol
PubChem CID 74594
IUPAC Name 2-hydroxy-3-[(2-methylprop-2-enoyl)oxy]propyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(O)COC(=O)C(C)=C

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 g
Formula Weight 228.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycerol Dimethacrylate (mixture of 1,2- and 1,3-form) (stabilized with MEHQ)
EINECSNumber (2)-1063&(2)-0958
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.