Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol Formal (mixture of 1,3-Dioxan-5-ol and 4-Hydroxymethyldioxolane) 98.0+%, TCI America™
SDP

Catalog No. G0244500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0244500ML 500 mL
G024425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0244500ML Supplier TCI America Supplier No. G0244500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99569-11-6
Molecular Formula C8H16O6
Molecular Weight (g/mol) 208.21
MDL Number MFCD00003218
InChI Key JIUMSISXCKWZTA-UHFFFAOYSA-N
PubChem CID 251541
IUPAC Name 1,3-dioxan-5-ol;1,3-dioxolan-4-ylmethanol
SMILES C1C(COCO1)O.C1C(OCO1)CO

Specifications

Color Yellow
Boiling Point 97°C
Quantity 500 mL
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycerol Formal (mixture of 1,3-Dioxan-5-ol and 4-Hydroxymethyldioxolane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.