Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol (Glycerin), ACS Reagent, Reagents

Catalog No. 01337946
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Glass Bottle
HDPE Winpak
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-946 500 mL Glass Bottle
01-337-949 20 L HDPE Winpak
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-946 Supplier Reagents Holdings Llc Supplier No. C1210500500C
Add to Cart
Add to Cart

Glycerol (Glycerin), ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym 1,2,3-Propanetriol, Glycerine
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18.2°C
Color Colorless
Boiling Point 544°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 1.2613
Grade ACS
Physical Form Liquid
Chemical Name or Material Glycerol (Glycerin), ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.