Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol Phosphate Disodium Salt Hydrate, Spectrum™ Chemical
SDP

Catalog No. 18602988 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-988 25 g Poly Bottle
18-602-989 100 g Poly Bottle
18-602-990 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-988 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G221625GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

G2216, 55073-41-1, C3H7Na2O6P·xH2O

Spectrum™ Chemical Glycerol Phosphate Disodium Salt Hydrate , also known as Glycerophosphate, is an organophosphate and a component of glycerophospholipids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 55073-41-1

Specifications

CAS Max % 100%
Grade Reagent
Quantity 25 g
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.