Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol Solution, 85%, Solstice

Catalog No. 60047006 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-047-006 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-006 Supplier Solstice Supplier No. 497831L

missing translation for 'validMainframeMsg'

Tested according to European Pharmacopoeia, 85%

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
Density 1.252g/mL at 25°C
pH Neutral
Boiling Point 290°C (1013 hPa)
Flash Point 160°C (320°F)
Odor Odorless
Packaging Glass bottle
Linear Formula HOCH2CH(OH)CH2OH · aq
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Solubility Information Completely miscible in water
Formula Weight 92.09g/mol (anhydrous basis)
Physical Form Liquid
Chemical Name or Material Glycerol solution, Tested according to Ph.Eur.
Show More Show Less
RTECSNumber 200-289-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.