Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Glycerophosphate disodium salt, pentahydrate, 99.4%, MP Biomedicals™

Catalog No. ICN19520691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19520691 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19520691 Supplier MP Biomedicals, Inc Supplier No. 0219520691

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

CAS 819-83-0
Color White
Assay Percent Range 0.994
Molecular Formula C3H7Na2O6P
MDL Number MFCD00002135 MFCD28134611 MFCD00149083 MFCD00150339 MFCD00002135
Synonym glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2o
InChI Key AVPCPPOOQICIRJ-UHFFFAOYSA-L
SMILES [Na+].[Na+].OCC(CO)OP([O-])([O-])=O
IUPAC Name disodium 2-(phosphonatooxy)propane-1,3-diol
Molecular Weight (g/mol) 216.04
PubChem CID 131860121
Formula Weight 306
Quantity 1 kg
Physical Form Powder
Chemical Name or Material β-Glycerophosphate disodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.