Learn More
Cytiva Glycine 2.0, 100mL
Catalog No. 501055299 Shop All Cytiva Products
Encompass
Aqueous buffer containing 10 mM glycine-HCl at pH 2.0
Catalog No. 50-105-5299 Supplier Cytiva Supplier No. BR100355
Aqueous buffer containing 10 mM glycine-HCl at pH 2.0