Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glycine, >99%, Calbiochem™,

Catalog No. 35701KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
35-701-KG 1 kg Glass Bottle
35-705-00GM 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-701-KG Supplier MilliporeSigma™ Supplier No. 35701KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Color White
Packaging Glass Bottle
Quantity 1 kg
Percent Purity >99% (Assay)
Physical Form Crystalline Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.