Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine Anhydride 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
1kg
Packaging:
Glass bottle
106-57-0
C4H6N2O2
114.1
MFCD00006009
BXRNXXXXHLBUKK-UHFFFAOYSA-N
2,5-piperazinedione,2,5-dioxopiperazine,2,5-diketopiperazine,cyclodiglycine,cycloglycylglycine,cyclo glycylglycyl,glycylglycine lactam,diglycolyl diamide,glycine, bimol. cyclic peptide,alpha,gamma-diacipiperazine
7817
CHEBI:16535
piperazine-2,5-dione
C1C(=O)NCC(=O)N1
106-57-0
C4H6N2O2
114.1
MFCD00006009
BXRNXXXXHLBUKK-UHFFFAOYSA-N
2,5-piperazinedione,2,5-dioxopiperazine,2,5-diketopiperazine,cyclodiglycine,cycloglycylglycine,cyclo glycylglycyl,glycylglycine lactam,diglycolyl diamide,glycine, bimol. cyclic peptide,alpha,gamma-diacipiperazine
7817
CHEBI:16535
piperazine-2,5-dione
C1C(=O)NCC(=O)N1
Packaging Glass bottle
Loss on Drying 0.5% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Quantity 25g
Melting Point >300.0°C
Color White to Yellow
Assay Percent Range 98%
Beilstein 24, 264
Merck Index 15, 7578
Solubility Information Solubility in water: 142g/L water (20 c). Other solubilities: soluble in hcl, slightly soluble in ethanol
Formula Weight 114.1
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Glycine anhydride, 98%
Show More Show Less

EINECSNumber : 203-411-5

RTECSNumber : TL6325250

TSCA : TSCA