Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, PlusOne™, Cytiva

Catalog No. 45000238 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-238 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-238 Supplier Cytiva Supplier No. 17132301
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use as a component of electrophoresis buffers. Glycine, PlusOne™, Cytiva is often a component of electrophoresis buffers, especially in the Laemmli discontinuous buffer system for SDS-PAGE.

  • Used in IEF and PAGE
  • Soluble in cold water and hot water, Slightly soluble in acetone
  • Insoluble in diethyl ether
  • Solid
  • Used as a component of buffers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6 , 7732-18-5
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Assay Percent Range ≥99.7%
Quantity 500 g
Formula Weight 75.07
Grade Electrophoresis
Chemical Name or Material Glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.