Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, Ultrapure, 99%, Spectrum™ Chemical
SDP

Catalog No. 18610746 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
12 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-746 1 kg Poly Bottle
18-610-749 5 kg Poly Bottle
18-610-748 12 kg Poly Bottle
18-610-747 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-746 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GL1571KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56-40-6

Specifications

Absorbance 0.1 (1 M Solution at 280 nm)
CAS Max % 1
Chloride 7e-05
Heavy Metals 2e-05
Identification To Pass Test
Loss on Drying 0.002
Packaging Poly Bottle
Sulfate 6.5e-05
Quantity 1 kg
Percent Purity 99%
Grade Ultrapure
Chemical Name or Material Glycine
Show More Show Less

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.