Learn More
Medchemexpress LLC HY-137179 10mg Medchemexpress, Glyco-diosgenin CAS:1402423-29-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13717910MG
Encompass_Preferred
Medchemexpress, HY-137179 10mg Glyco-diosgenin CAS:1402423-29-3 Glyco-diosgenin is a synthetic surfactant and detergent for extracting proteins from membranes for structure and function studies, and single-particle cryo-electron microscopy (cryoEM) studies of membrane proteins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1473
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.