Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycol chitosan, 76.2%, MP Biomedicals™

Catalog No. icn15724691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15724691 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15724691 Supplier MP Biomedicals, Inc Supplier No. 0215724691

Glycol Chitosan, Light Yellow Powder

  • Glycol chitosan increases membrane permeability and leakage in Glycine max Harosoy 63w cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39280-86-9
Molecular Formula (C8H15NO5)n

Specifications

Color Tan to White
Assay Percent Range 76.2%
Quantity 1 g
Synonym Glycol chitin, deacetylated
Formula Weight (205.22)n
Physical Form Powder
Chemical Name or Material Glycol chitosan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.