Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycolaldehyde dimer

Catalog No. G68051G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G68051G 1 g
G68055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G68051G Supplier Sigma Aldrich Supplier No. G68051G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Glycolaldehyde dimer may be used in the synthesis of 3,4-diaza-2-hexene-1,6-diol, which can undergo hydrogenation to form 1,2-bis(2-hydroxyethyl)hydrazine
  • It undergoes cycloaddition with 2,3-dihydrofuran in the presence of a chiral catalyst to form fused bicyclic tetrahydrofuran (bis-thf) alcohol, a key moiety of hiv protease inhibitors

Specifications

CAS 23147-58-2
Molecular Formula C4H8O4
Linear Formula C4H8O4
MDL Number MFCD00012133
Quantity 1 g
Synonym 1,4-Dioxane-2,5-diol; 2,5-Dihydroxy-1,4-dioxane; Hydroxyacetaldehyde dimer
Molecular Weight (g/mol) 120.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.