Learn More
Sigma Organic Chemistry Glycolic acid solution | 100ml | 79-14-1 | MFCD00004312
Supplier: Sigma Organic Chemistry 420581100ML
Glycolic acid solution , 100ml
About this item:
CAS #: 79-14-1
MDL #: MFCD00004312
Purity: 63-72%
Molecular Weight: 76.05
UNSPSC Code: 12352100
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.