Learn More
Sigma Organic Chemistry Glycolic acid solution | 1L | 79-14-1 | MFCD00004312
Supplier: Sigma Organic Chemistry 4206031L
Glycolic acid solution , 1L
About this item:
CAS #: 79-14-1
MDL #: MFCD00004312
Molecular Weight: 76.05
UNSPSC Code: 12352100
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.