Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycolide 98.0+%, TCI America™
SDP

Catalog No. G046725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G046725G 25 g
G04675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G046725G Supplier TCI America Supplier No. G046725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 502-97-6
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00081108
InChI Key RKDVKSZUMVYZHH-UHFFFAOYSA-N
Synonym glycolide, glycollide, unii-yrz676pgu6, p-dioxane-2,5-dione, yrz676pgu6, acmc-1aybf, 1,4-dioxan-2,5-dione, 2,5-dioxo-1,4-dioxane, p-dioxane-2,5-dione 8ci
PubChem CID 65432
IUPAC Name 1,4-dioxane-2,5-dione
SMILES O=C1COC(=O)CO1

Specifications

Melting Point 84°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 130°C
Formula Weight 116.07
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Glycolide
EINECSNumber (5)-6815&(5)-6865
TSCA Yes
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.