Learn More
ACROBiosystems Recombinant Protein;Mouse Glycophorin A / CD235a Protein, His Tag & Fc Tag;HEK293;50UG;CDA,CD235a,Glycophorin-A,Gypa
Supplier: ACROBiosystems CDAM526X50UG
Recombinant Protein;Mouse Glycophorin A / CD235a Protein, His Tag & Fc Tag;50UG;Mouse CD235a, His Tag, Fc Tag (CDA-M526x) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Val 108 (Accession # P14220).;This protein carries a human IgG1 Fc tag at the N-terminus and a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.6 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.