Learn More
ACROBiosystems Glycoprotein D HSV-2 100ug
Supplier:  ACROBiosystems GLDY111100UG
Encompass_Preferred
Herpesvirus infections are widely spread throughout the world population. Herpes simplex virus (HSV) belongs to the α-herpesvirus subfamily. There are two main types of HSV HSV-1 and HSV-2 which infect humans. HSV-2 mainly causes genital lesions whereas HSV-1 is involved in both oral and genital infections. Glycoprotein D (gD) is a structural component of the herpes simplex virus type 1 (HSV-1) envelope which is essential for virus entry and fusion with host cells. gD plays an important role by binding to the host receptors such as herpes virus entry mediator (HVEM) and nectin-1 a member of the immunoglobulin (Ig)-like cell adhesion molecules.
Catalog No. 50-253-5009
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.