Learn More
ACROBiosystems Varicella Zoster Virus Glycoprotein E (VZV gE) ELISA Kit
Supplier:  ACROBiosystems RASA10296TESTS
Encompass_Preferred
Varicella-zoster virus (VZV) the etiologic agent of chickenpox and herpes zoster [HZ], is highly contagious and still endemic worldwide. Glycoprotein E (gE) is one of the known glycoproteins (gB, gC, gE, gH, gI, gK, gI) of VZV that is most abundantly expressed on the surface of virus and infected cells, playing an important role in viral replication and cell-to-cell spread. The strongly immunogenic gE can provide strong IgG signal in body fluid, which makes it ideal to be developed as an antigen for analysis of Immunogenicity in the development of VZV vaccine.Therefore,It's helpful to develop the Varicella Zoster Virus Glycoprotein E (VZV gE) ELISA Kit to quantitative detection the VZV gE antigen in vaccine samples during the manufacture and quality control of vaccine development.
Catalog No. 50-233-1740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.