Description
Includes
NS0-expressed hr-ENTPD3/CD39L3, UDP, Phosphatase Buffer 1, 100mMMnCl2, Phosphate Standard, Malachite Green Reagents A and B
Uses the nucleotidase ENTPD3/CD39L3 as a coupling phosphatase to remove inorganic phosphate quantitatively from the leaving nucleotide diphosphate, such as UDP or GDP, generated during glycosyltransferase reactions
No time-consuming separation steps , as
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Kit |