Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Glycylglycylglycine, 98+%

Catalog No. AAA1377803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1377803 1 g
AAA1377806 5 g
AAA1377814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1377803 Supplier Thermo Scientific Chemicals Supplier No. A1377803
Add to Cart

CAS: 556-33-2 | C6H11N3O4 | 189.17 g/mol

Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases

Solubility
Soluble in water (94.6 mg/ml at 20°C), methanol (50 mg/ml), and 80% acetic acid (50 mg/ml).

Notes
Store at -20°C. Protect from moisture. Avoid excessive heat and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-33-2
Molecular Formula C6H11N3O4
Molecular Weight (g/mol) 189.17
MDL Number MFCD00036223
InChI Key XKUKSGPZAADMRA-UHFFFAOYSA-N
Synonym triglycine, glycylglycylglycine, glycyl-glycyl-glycine, diglycylglycine, gly-gly-gly, h-gly-gly-gly-oh, n-n-glycylglycyl glycine, glyglygly, glycine, glycylglycyl, unii-cvk73zdq8b
PubChem CID 11161
ChEBI CHEBI:63961
IUPAC Name 2-[[2-[(2-aminoacetyl)amino]acetyl]amino]acetic acid
SMILES NCC(=O)NCC(=O)NCC(O)=O

Specifications

Melting Point ∼240°C (decomposition)
Beilstein 1711130
Merck Index 14,6579
Solubility Information Soluble in water (94.6mg/ml at 20°C),methanol (50mg/ml),and 80% acetic acid (50mg/ml).
Quantity 1 g
Formula Weight 191.14
Percent Purity 99%
Chemical Name or Material Glycylglycylglycine
EINECSNumber 209-122-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.