Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycylsarcosine 97.0+%, TCI America™
SDP

Catalog No. G01381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01381G 1g
G01385G 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G01381G Supplier TCI America Supplier No. G01381G
Add to Cart
Add to Cart

Specifications

Purity ≥97.0% (HPLC)
CAS 29816-01-1
Color White
Content And Storage Refrigerator
Format Crystal-Powder at 20°C
Molecular Weight (g/mol) 146.15
Product Type Glycylsarcosine
Quantity 1g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.